missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-leucine, 98%

CAS: 114360-54-2 | C21H23NO4 | 353.42 g/mol

Supplier:  Thermo Scientific Chemicals H2904106

 View more versions of this product

Catalog No. AAH2904106


Only null left
Explore available promotions

Description

Description

A convergent synthesis of the proposed structure of (+)-pestalazine B has been achieved in 4 steps using the N-alkylation of an unprotected tryptophan diketopiperazine with a 3a-bromopyrrolidinoindoline where the condensation between L-tryptophan methyl ester and N-Fmoc-D-leucine in the presence of EDC as coupling agent afforded the corresponding L-Trp-D-Leu dipeptide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A convergent synthesis of the proposed structure of (+)-pestalazine B has been achieved in 4 steps using the N-alkylation of an unprotected tryptophan diketopiperazine with a 3a-bromopyrrolidinoindoline where the condensation between L-tryptophan methyl ester and N-Fmoc-D-leucine in the presence of EDC as coupling agent afforded the corresponding L-Trp-D-Leu dipeptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Specifications

Specifications

N-Fmoc-D-leucine
151°C to 154°C
MFCD00062957
fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid
CBPJQFCAFFNICX-TWYLJJHKNA-N
CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
353.42
353.42
114360-54-2
C21H23NO4
5 g
Insoluble in water.
+25° (c=1 in DMF)
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
1549477
98%

Chemical Identifiers

114360-54-2
353.42
CBPJQFCAFFNICX-TWYLJJHKNA-N
1549477
CC(C)C[C@@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O
C21H23NO4
MFCD00062957
fmoc-d-leu-oh, fmoc-d-leucine, n-fmoc-d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl-d-leucine, n-9-fluorenylmethyloxycarbonyl-d-leucine, n-9-fluorenylmethoxycarbonyl-l-leucine, d-leucine, n-9h-fluoren-9-ylmethoxy carbonyl, n-alpha-9-fluorenylmethyloxycarbonyl-d-leucine, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methyl-pentanoic acid, 2r-2-9h-fluoren-9-ylmethoxycarbonylamino-4-methylpentanoic acid
(2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only