missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-aspartic acid 4-tert-butyl ester, 98%

Catalog No. AAH6235606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6235606 5 g
AAH6235603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6235606 Supplier Thermo Scientific Chemicals Supplier No. H6235606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 112883-39-3 | C23H25NO6 | 411.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112883-39-3
Molecular Formula C23H25NO6
Molecular Weight (g/mol) 411.45
MDL Number MFCD00077050
InChI Key FODJWPHPWBKDON-UHFFFAOYNA-N
Synonym fmoc-d-asp otbu-oh, r-2-9h-fluoren-9-yl methoxy carbonyl amino-4-tert-butoxy-4-oxobutanoic acid, fmoc-d-asp obut-oh, fmoc-d-aspartic acid beta-tert-butyl ester, n-alpha-fmoc-d-aspartic acid beta-t-butyl ester, 2r-4-tert-butoxy-2-9h-fluoren-9-ylmethoxy carbonyl amino-4-oxobutanoic acid, fmoc-dasp otbu, ambotzfaa1309, fmoc-dasp otbu-oh, pubchem10009
PubChem CID 7018816
SMILES CC(C)(C)OC(=O)CC(NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Quantity 5 g
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
Formula Weight 411.45
Percent Purity 98%
Chemical Name or Material N-Fmoc-D-aspartic acid 4-tert-butyl ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only