missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-allo-isoleucine, 98%

CAS: 118904-37-3 | C21H23NO4 | 353.42 g/mol

$347.77 - $1514.63

Chemical Identifiers

CAS 118904-37-3
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD00077062
InChI Key QXVFEIPAZSXRGM-KINVLMLONA-N
Synonym fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
PubChem CID 6992517
IUPAC Name (2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid
SMILES CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6345503
View Documents
Thermo Scientific Chemicals
H6345503
1 g
Each for $347.77
Only null left
 
AAH6345506
View Documents
Thermo Scientific Chemicals
H6345506
5 g
Each for $1,514.63
Only null left
 
Description

Description

N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-D-allo-isoleucine used as a intermediate in pharmaceutical and chemical synthesis.

Solubility
Slightly soluble in water.

Notes
Refrigerate at -20°C. Keep container tightly closed. Store away from oxidizing agent.
Specifications

Chemical Identifiers

118904-37-3
353.42
QXVFEIPAZSXRGM-KINVLMLONA-N
6992517
CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
C21H23NO4
MFCD00077062
fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid

Specifications

118904-37-3
MFCD00077062
Slightly soluble in water.
CC[C@H](C)[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O
1 g
6992517
98%
N-Fmoc-D-allo-isoleucine
C21H23NO4
fmoc-d-allo-ile-oh, fmoc-d-allo-isoleucine, fmoc-d-allo-lle-oh, 2r,3s-n-fmoc-2-amino-3-methylpentanoic acid, 2r,3s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylpentanoic acid, d-alloisoleucine, n-9h-fluoren-9-ylmethoxy carbonyl, 2r,3s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-methylpentanoic acid, ambotzfaa1328, pubchem15619
QXVFEIPAZSXRGM-KINVLMLONA-N
(2R,3S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylpentanoic acid
353.42
353.41
Solid
Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

RUO – Research Use Only