missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-nitro-L-phenylalanine, 98%

Catalog No. p-7034291
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5196203 1 g
AAH5196206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5196203 Supplier Thermo Scientific Chemicals Supplier No. H5196203
Only null left

CAS: 95753-55-2 | C24H20N2O6 | 432.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 95753-55-2
Molecular Formula C24H20N2O6
Molecular Weight (g/mol) 432.43
MDL Number MFCD00057810
InChI Key RZRRJPNDKJOLHI-QFIPXVFZSA-N
Synonym fmoc-4-nitro-l-phenylalanine, fmoc-phe 4-no2-oh, fmoc-l-4-no2-phe-oh, fmoc-p-nitro-phe-oh, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-nitrophenyl propanoic acid, n-fmoc-4-nitro-l-phenylalanine, n-9h-fluoren-9-ylmethoxy carbonyl-4-nitro-l-phenylalanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-4-nitrophenyl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino-3-4-nitrophenyl propanoic acid, fmoc-phe no2-oh
PubChem CID 7016054
IUPAC Name (2S)-2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(4-nitrophenyl)propanoic acid
SMILES OC(=O)[C@H](CC1=CC=C(C=C1)[N+]([O-])=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 213°C to 223°C
Quantity 1 g
Beilstein 5178656
Formula Weight 432.43
Percent Purity 98%
Chemical Name or Material N-Fmoc-4-nitro-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only