missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-methoxy-L-phenylalanine, 95%

Catalog No. p-7034432
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5211703 1 g
AAH5211706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5211703 Supplier Thermo Scientific Chemicals Supplier No. H5211703
Only null left

CAS: 77128-72-4 | C25H23NO5 | 417.461 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 77128-72-4
Molecular Formula C25H23NO5
Molecular Weight (g/mol) 417.461
MDL Number MFCD00153368
InChI Key JYQODLWFOPCSCS-QHCPKHFHSA-N
Synonym fmoc-tyr me-oh, fmoc-o-methyl-l-tyrosine, fmoc-4-methoxy-l-phenylalanine, fmoc-l-4-methoxyphenylalanine, fmoc-4-methoxy-phe-oh, fmoc-o-methyl-tyr-oh, fmoc-l-tyr me, fmoc-4-methoxy-l-phe-oh, fmoc-l-4-methoxyphe
PubChem CID 2762281
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(4-methoxyphenyl)propanoic acid
SMILES COC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Formula Weight 417.45
Percent Purity 95%
Chemical Name or Material N-Fmoc-4-methoxy-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only