missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-4-chloro-L-phenylalanine, 98%

Catalog No. AAH6344806
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6344806 5 g
AAH6344803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6344806 Supplier Thermo Scientific Chemicals Supplier No. H6344806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

missing translation for 'validMainframeMsg'

CAS: 175453-08-4 | C24H20ClNO4 | 421.877 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 175453-08-4
Molecular Formula C24H20ClNO4
Molecular Weight (g/mol) 421.877
MDL Number MFCD00080281
InChI Key CQPNKLNINBUUOM-QFIPXVFZSA-N
Synonym fmoc-4-chloro-l-phenylalanine, s-n-fmoc-4-chlorophenylalanine, fmoc-phe 4-cl-oh, fmoc-l-4-chlorophenylalanine, fmoc-l-phe 4-cl-oh, fmoc-4-chloro-phe-oh, fmoc-l-4-chlorophe, 2s-3-4-chlorophenyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-chlorophenyl propanoic acid, phenylalanine, 4-chloro-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 2734471
IUPAC Name (2S)-3-(4-chlorophenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CC=C(C=C4)Cl)C(=O)O

Specifications

Beilstein 9437838
Quantity 5 g
Formula Weight 421.87
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-4-chloro-L-phenylalanine
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only