missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-4-benzoyl-D-phenylalanine, 98%, Thermo Scientific Chemicals
$494.29 - $494.29
Chemical Identifiers
| CAS | 117666-97-4 |
|---|---|
| Molecular Formula | C31H25NO5 |
| Molecular Weight (g/mol) | 491.543 |
| MDL Number | MFCD00237666 |
| InChI Key | SYOBJKCXNRQOGA-MUUNZHRXSA-N |
| Synonym | fmoc-d-bpa-oh, fmoc-4-benzoyl-d-phenylalanine, fmoc-d-4-benzoylphenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid, 2r-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1665, boc-p-bz-d-phe-oh, fmoc-p-bz-d-phe-oh, fmoc-d-bpa-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-4-benzoyl-d-phenylalanine |
| PubChem CID | 14233361 |
| IUPAC Name | (2R)-3-(4-benzoylphenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
| SMILES | C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6603006
|
Thermo Scientific Chemicals
H6603006 |
5 g |
Each for $494.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 117666-97-4 | |
| 491.543 | |
| SYOBJKCXNRQOGA-MUUNZHRXSA-N | |
| 14233361 | |
| C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35 |
| C31H25NO5 | |
| MFCD00237666 | |
| fmoc-d-bpa-oh, fmoc-4-benzoyl-d-phenylalanine, fmoc-d-4-benzoylphenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid, 2r-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1665, boc-p-bz-d-phe-oh, fmoc-p-bz-d-phe-oh, fmoc-d-bpa-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-4-benzoyl-d-phenylalanine | |
| (2R)-3-(4-benzoylphenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid |
Specifications
| 117666-97-4 | |
| C31H25NO5 | |
| fmoc-d-bpa-oh, fmoc-4-benzoyl-d-phenylalanine, fmoc-d-4-benzoylphenylalanine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-benzoylphenyl propanoic acid, 2r-3-4-benzoylphenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, ambotzfaa1665, boc-p-bz-d-phe-oh, fmoc-p-bz-d-phe-oh, fmoc-d-bpa-oh hplc, n-alpha-9-fluorenylmethyloxycarbonyl-4-benzoyl-d-phenylalanine | |
| C1=CC=C(C=C1)C(=O)C2=CC=C(C=C2)CC(C(=O)O)NC(=O)OCC3C4=CC=CC=C4C5=CC=CC=C35 | |
| 5 g | |
| 14233361 | |
| 98% | |
| N-Fmoc-4-benzoyl-D-phenylalanine |
| Brown | |
| MFCD00237666 | |
| SYOBJKCXNRQOGA-MUUNZHRXSA-N | |
| (2R)-3-(4-benzoylphenyl)-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid | |
| 491.543 | |
| 491.54 | |
| Powder |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only