missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-cyclohexyl-L-alanine, 98%

CAS: 135673-97-1 | C24H27NO4 | 393.48 g/mol

Supplier:  Thermo Scientific Chemicals H6391003

 View more versions of this product

Catalog No. AAH6391003


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Fmoc-3-cyclohexyl-L-alanine
C24H27NO4
7052264
HIJAUEZBPWTKIV-UHFFFAOYNA-N
(2S)-3-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
393.48
393.48
Solid
135673-97-1
MFCD00065614
fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
1 g
978326
98%

Chemical Identifiers

135673-97-1
393.48
HIJAUEZBPWTKIV-UHFFFAOYNA-N
978326
OC(=O)C(CC1CCCCC1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12
C24H27NO4
MFCD00065614
fmoc-cha-oh, fmoc-3-cyclohexyl-l-alanine, fmoc-l-cyclohexylalanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-cyclohexylpropanoic acid, n-fmoc-3-cyclohexyl-l-alanine, n-9h-fluoren-9-ylmethoxy carbonyl-3-cyclohexyl-l-alanine, n-alpha-9-fluorenylmethyloxycarbonyl-l-cyclohexylalanine, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxycarbonylamino propanoic acid, 2s-3-cyclohexyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-3-cyclohexylalanine
(2S)-3-cyclohexyl-2-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only