missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-3-(1-naphthyl)-D-alanine, 98%
CAS: 138774-93-3 | C28H23NO4 | 437.50 g/mol
$99.22 - $331.88
Chemical Identifiers
| CAS | 138774-93-3 |
|---|---|
| Molecular Formula | C28H23NO4 |
| Molecular Weight (g/mol) | 437.50 |
| MDL Number | MFCD00191200 |
| InChI Key | ORWNVJDLEMVDLV-UHFFFAOYNA-N |
| Synonym | fmoc-3-1-naphthyl-d-alanine, fmoc-d-1-nal-oh, fmoc-d-1-naphthylalanine, fmoc-beta-1-naphthyl-d-alanine, fmoc-d-3-1-naphthyl-alanine, fmoc-3-1-naphthyl-d-ala, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-naphthalen-1-yl-propionic acid, n-fmoc-3-1-naphthyl-l-alanine, ambotzfaa1659 |
| PubChem CID | 7129831 |
| SMILES | OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH6368803
|
Thermo Scientific Chemicals
H6368803 |
1 g |
Each for $99.22
|
|
|||||
|
AAH6368806
|
Thermo Scientific Chemicals
H6368806 |
5 g |
Each for $331.88
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 138774-93-3 | |
| 437.50 | |
| ORWNVJDLEMVDLV-UHFFFAOYNA-N | |
| 7129831 |
| C28H23NO4 | |
| MFCD00191200 | |
| fmoc-3-1-naphthyl-d-alanine, fmoc-d-1-nal-oh, fmoc-d-1-naphthylalanine, fmoc-beta-1-naphthyl-d-alanine, fmoc-d-3-1-naphthyl-alanine, fmoc-3-1-naphthyl-d-ala, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-naphthalen-1-yl-propionic acid, n-fmoc-3-1-naphthyl-l-alanine, ambotzfaa1659 | |
| OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 |
Specifications
| 138774-93-3 | |
| MFCD00191200 | |
| fmoc-3-1-naphthyl-d-alanine, fmoc-d-1-nal-oh, fmoc-d-1-naphthylalanine, fmoc-beta-1-naphthyl-d-alanine, fmoc-d-3-1-naphthyl-alanine, fmoc-3-1-naphthyl-d-ala, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, r-2-9h-fluoren-9-ylmethoxycarbonylamino-3-naphthalen-1-yl-propionic acid, n-fmoc-3-1-naphthyl-l-alanine, ambotzfaa1659 | |
| OC(=O)C(CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12 | |
| 1 g | |
| 7129831 | |
| 98% | |
| N-Fmoc-3-(1-naphthyl)-D-alanine |
| C28H23NO4 | |
| 9160026 | |
| ORWNVJDLEMVDLV-UHFFFAOYNA-N | |
| 2-({[(9H-fluoren-9-yl)methoxy]carbonyl}amino)-3-(naphthalen-1-yl)propanoic acid | |
| 437.50 | |
| 437.49 | |
| Solid |
Safety and Handling
TSCA : No
storageNote1 : 2° to 8°C
Recommended Storage : Keep cold
RUO – Research Use Only