missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-2-octyl-L-glycine, 95%

CAS: 193885-59-5 | C25H31NO4 | 409.526 g/mol

$174.05 - $699.05

Chemical Identifiers

CAS 193885-59-5
Molecular Formula C25H31NO4
Molecular Weight (g/mol) 409.526
MDL Number MFCD01311753
InChI Key LSMLSLHVRMSYPT-QHCPKHFHSA-N
Synonym fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
PubChem CID 23257324
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
SMILES CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH6603103
View Documents
Thermo Scientific Chemicals
H6603103
1 g
Each for $174.05
Only null left
 
AAH6603106
View Documents
Thermo Scientific Chemicals
H6603106
5 g
Each for $699.05
Only null left
 
Description

Description

A homolog of lipophilic building block for SAR studies.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A homolog of lipophilic building block for SAR studies.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Specifications

Chemical Identifiers

193885-59-5
409.526
LSMLSLHVRMSYPT-QHCPKHFHSA-N
23257324
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
C25H31NO4
MFCD01311753
fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid

Specifications

140°C to 143°C
White
MFCD01311753
Slightly soluble in water.
CCCCCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13
1 g
23257324
98%
N-Fmoc-2-octyl-L-glycine
193885-59-5
C25H31NO4
fmoc-octyl-gly-oh, fmoc-2s-2-amino-decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid, ambotzfaa1598, fmoc-s-octylglycine, s-n-fmoc-octylglycine-oh, n-alpha-9-fluorenylmethyloxycarbonyl-l-octylglycine, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino decanoic acid, s-2-9h-fluoren-9-yl methoxy carbonylamino decanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino decanoic acid
LSMLSLHVRMSYPT-QHCPKHFHSA-N
(2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)decanoic acid
409.526
409.53
Powder
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only