missing translation for 'onlineSavingsMsg'
Learn More

Ergodyne™ N-Ferno™ 6466 Thermal Jacket

Catalog No. p-7163179
Change view
Click to view available options
Size:
Small
Medium
Large
2X-Large
3X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19910188 Small
19910189 Medium
19910190 Large
19910192 2X-Large
19910193 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19910188 Supplier Ergodyne™ Supplier No. 41112
Only null left

Rugged 500D shell repels water and protects users from the wind and cold while remaining breathable for all day comfort.

For use with: Roof Bolting, Mining, Oil/Gas, Demolition, Heavy Construction, Ironwork, Mechanics

Specifications

Size Small
Material Nylon (500D)
Color Black