missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmorpholine, 99%

Catalog No. p-130998
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC149401000 100 mL Glass bottle
AC149405000 500 mL Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC149401000 Supplier Thermo Scientific Chemicals Supplier No. 149401000
Only null left

CAS: 100-74-3 | C6H13NO | 115.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 100-74-3
Molecular Formula C6H13NO
Molecular Weight (g/mol) 115.17
MDL Number MFCD00006177
InChI Key HVCNXQOWACZAFN-UHFFFAOYSA-N
Synonym n-ethylmorpholine, morpholine, 4-ethyl, ethylmorpholine, n-ethylmorfolin, n-ethyl morpholine, n-ethylmorfolin czech, unii-ecm0g991fq, n-ethyl-morpholine, ccris 4818, ecm0g991fq
PubChem CID 7525
IUPAC Name 4-ethylmorpholine
SMILES CCN1CCOCC1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -63.0°C
Color Brown-Gray to Cream
Density 0.9000g/mL
Boiling Point 139.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.4400 to 1.4420
Quantity 100 mL
Beilstein 27, 6
Fieser 01,383
Specific Gravity 0.9
Solubility Information Solubility in water: miscible with water. Other solubilities: soluble in alcohol,ether,acetone and benzene
Formula Weight 115.17
Percent Purity 99%
Physical Form Powder
Chemical Name or Material N-Ethylmorpholine, 99%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Flammable liquid and vapor.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 202-885-0
RTECSNumber QE4025000
TSCA TSCA

RUO – Research Use Only