missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmethylamine, 97%

Catalog No. p-3757885
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC314680250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC314680250 Supplier Thermo Scientific Chemicals Supplier No. 314680250

CAS: 624-78-2 | C3H9N | 59.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 624-78-2
Molecular Formula C3H9N
Molecular Weight (g/mol) 59.11
MDL Number MFCD00009030
InChI Key LIWAQLJGPBVORC-UHFFFAOYSA-N
Synonym n-ethylmethylamine, methylethylamine, ethanamine, n-methyl, n-methylethylamine, ethyl methyl amine, ethylmethylamine, nmea, n-methyl-n-ethylamine, ethylamine, n-methyl, ethyl-methyl-amine
PubChem CID 12219
IUPAC Name N-methylethanamine
SMILES CCNC

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 0.6800g/mL
Boiling Point 33.0°C to 34.0°C
Flash Point <−34°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Refractive Index 1.3730 to 1.3770
Linear Formula C2H5NHCH3
Quantity 25 g
Specific Gravity 0.68
Solubility Information Solubility in water: soluble
Formula Weight 59.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Ethylmethylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Extremely flammable liquid and vapor.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful in contact with skin.
Harmful if swallowed.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse
EINECSNumber 210-862-1

RUO – Research Use Only