missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide 98.0+%, TCI America™

Supplier:  TCI America E013625G

 View more versions of this product

Catalog No. E013625G

Only null left
Explore available promotions

Description

Description

Specifications

Specifications

N-Ethylmaleimide
46°C
210°C
MFCD00005509
2928
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
Crystalline Powder
128-53-0
White
C6H7NO2
25 g
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CCN1C(=O)C=CC1=O
125.127
CHEBI:44485
≥98.0% (HPLC,N)

Chemical Identifiers

128-53-0
125.127
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione
C6H7NO2
MFCD00005509
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CHEBI:44485
CCN1C(=O)C=CC1=O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : UX9625000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions