missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide 98.0+%, TCI America™

Catalog No. E01361G
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01361G 1 g
E01365G 5 g
E013625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. E01361G Supplier TCI America Supplier No. E01361G

Chemical Identifiers

CAS 128-53-0
Molecular Formula C6H7NO2
Molecular Weight (g/mol) 125.127
MDL Number MFCD00005509
InChI Key HDFGOPSGAURCEO-UHFFFAOYSA-N
Synonym n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
PubChem CID 4362
ChEBI CHEBI:44485
IUPAC Name 1-ethylpyrrole-2,5-dione
SMILES CCN1C(=O)C=CC1=O

Specifications

Melting Point 46°C
Color White
Boiling Point 210°C
Quantity 1 g
UN Number 2928
Formula Weight 125.13
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethylmaleimide
RTECSNumber UX9625000
TSCA Yes
Recommended Storage Refrigerator