Learn More
N-Ethylmaleimide, 98+%
CAS: 128-53-0 | C6H7NO2 | 125.13 g/mol
Supplier: Thermo Scientific Chemicals 156100500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Ethylmaleimide | |
| 41.0°C to 45.0°C | |
| 210.0°C | |
| Authentic | |
| Glass bottle | |
| 50 g | |
| 15,3877 | |
| Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C) | |
| CCN1C(=O)C=CC1=O | |
| 125.13 | |
| CHEBI:44485 | |
| 99+% |
| 128-53-0 | |
| White | |
| 73°C | |
| 99% min. (GC) | |
| C6H7NO2 | |
| 21,IV,4629 | |
| n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide | |
| HDFGOPSGAURCEO-UHFFFAOYSA-N | |
| 1-ethylpyrrole-2,5-dione | |
| 4362 | |
| 125.13 | |
| Crystalline Powder |
Chemical Identifiers
| 128-53-0 | |
| 125.13 | |
| n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide | |
| CHEBI:44485 | |
| CCN1C(=O)C=CC1=O |
| C6H7NO2 | |
| HDFGOPSGAURCEO-UHFFFAOYSA-N | |
| 4362 | |
| 1-ethylpyrrole-2,5-dione |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
EINECSNumber : 204-892-4
RUO – Research Use Only