missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylmaleimide, 98+%

CAS: 128-53-0 | C6H7NO2 | 125.13 g/mol

Supplier:  Thermo Scientific Chemicals 156100100

 View more versions of this product

Catalog No. AC156100100


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Ethylmaleimide
41.0°C to 45.0°C
210.0°C
Authentic
Glass bottle
10 g
15,3877
Solubility in water: soluble. Other solubilities: 1115g/L toluene (20°C)
CCN1C(=O)C=CC1=O
125.13
CHEBI:44485
99+%
128-53-0
White
73°C
99% min. (GC)
C6H7NO2
21,IV,4629
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
HDFGOPSGAURCEO-UHFFFAOYSA-N
1-ethylpyrrole-2,5-dione
4362
125.13
Crystalline Powder

Chemical Identifiers

128-53-0
125.13
n-ethylmaleimide, ethylmaleimide, 1-ethyl-1h-pyrrole-2,5-dione, maleimide, n-ethyl, 1h-pyrrole-2,5-dione, 1-ethyl, maleic acid n-ethylimide, nem, usaf b-121, unii-o3c74acm9v, maleic acid-n-ethylimide
CHEBI:44485
CCN1C(=O)C=CC1=O
C6H7NO2
HDFGOPSGAURCEO-UHFFFAOYSA-N
4362
1-ethylpyrrole-2,5-dione
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Fatal if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 204-892-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only