missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylisopropylamine, 98%

CAS: 19961-27-4 | C5H13N | 87.17 g/mol

Supplier:  Thermo Scientific Chemicals L0408906

 View more versions of this product

Catalog No. AAL0408906


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

N-Ethylisopropylamine
0.72
16°C (60°F)
1.392
5 g
505952
RIVIDPPYRINTTH-UHFFFAOYSA-N
N-ethylpropan-2-amine
88318
98%
19961-27-4
71°C to 73°C
C5H13N
MFCD00015105
UN2733
n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine
CCNC(C)C
87.17
87.17

Chemical Identifiers

19961-27-4
87.17
RIVIDPPYRINTTH-UHFFFAOYSA-N
88318
C5H13N
MFCD00015105
n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine
CCNC(C)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.

P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c

H225-H302+H312+H332-H314

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.

EINECSNumber : 243-442-1

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only