missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylisopropylamine, 98%

Catalog No. AAL0408903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0408903 1 g
AAL0408906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0408903 Supplier Thermo Scientific Chemicals Supplier No. L0408903

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 19961-27-4 | C5H13N | 87.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 19961-27-4
Molecular Formula C5H13N
Molecular Weight (g/mol) 87.17
MDL Number MFCD00015105
InChI Key RIVIDPPYRINTTH-UHFFFAOYSA-N
Synonym n-ethylisopropylamine, ethylisopropylamine, 2-propanamine, n-ethyl, ethyl propan-2-yl amine, diethylamine, 1-methyl, isopropylamine, n-ethyl, n-ethyl-2-propanamine, n-ethyl-n-isopropylamine, isopropylethylamine, ethyl isopropylamine
PubChem CID 88318
SMILES CCNC(C)C

Specifications

Density 0.72
Boiling Point 71°C to 73°C
Flash Point 16°C (60°F)
Refractive Index 1.392
Quantity 1 g
UN Number UN2733
Beilstein 505952
IUPAC Name ethyl(propan-2-yl)amine
Formula Weight 87.17
Percent Purity 98%
Chemical Name or Material N-Ethylisopropylamine
Show More Show Less
Hazard Category H225-H302+H312+H332-H314
Hazard Statement GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P210-P233-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
EINECSNumber 243-442-1
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only