missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylheptylamine 98.0+%, TCI America™

Catalog No. E07805G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07805G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E07805G Supplier TCI America Supplier No. E07805G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 66793-76-8
Molecular Formula C9H21N
Molecular Weight (g/mol) 143.27
MDL Number MFCD00049012
InChI Key IUZZLNVABCISOI-UHFFFAOYSA-N
Synonym n-ethylheptylamine, n-ethyl-1-heptanamine, 1-heptanamine, n-ethyl, ethylheptylamine, ethyl heptyl amine, ethylheptyl-amine, n-ethylheptanamine, n-ethyl-n-heptylamine, n-heptyl-n-ethylamine, acmc-209nwl
PubChem CID 48038
IUPAC Name ethyl(heptyl)amine
SMILES CCCCCCCNCC

Specifications

Color Colorless
Boiling Point 181°C
Quantity 5 g
UN Number 2733
Formula Weight 143.27
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material N-Ethylheptylamine