missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylguanidine sulfate, 98%

Catalog No. aab2131506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2131506 5 g
AAB2131514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAB2131506 Supplier Thermo Scientific Chemicals Supplier No. B2131506
Get Up to $1,800 in Chemicals with Purchase
View all promotions
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 3482-86-8 | [CH3CH2NHC(=NH)NH2]2·H2SO4

1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Ethylguanidine sulfate may be used in chemical synthesis and thermal analysis studies. 1-Ethylguanidine sulfate is used as a reactant in the sunthesis of 3-Ethyl-6-methyl-isocytosines.

Solubility
Soluble in water.

Notes
Hygroscopic. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents,water, moisture.

Chemical Identifiers

CAS 3482-86-8
Molecular Formula [CH3CH2NHC(=NH)NH2]2·H2SO4
MDL Number MFCD00013130

Specifications

Melting Point ∼245°C (decomposition)
Odor Odorless
Quantity 5 g
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 272.33
Percent Purity 98%
Chemical Name or Material N-Ethylguanidine sulfate
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 222-467-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only