missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylethylenediamine, 98+%, Thermo Scientific™

$277.73

Chemical Identifiers

CAS 110-72-5
Molecular Formula C4H12N2
Molecular Weight (g/mol) 88.15
MDL Number MFCD00008166
InChI Key SCZVXVGZMZRGRU-UHFFFAOYSA-N
Synonym n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
PubChem CID 66071
IUPAC Name N'-ethylethane-1,2-diamine
SMILES CCNCCN
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC155550500
View Documents Promotion Details
N/A 50g Glass bottle
Each for $277.73
 
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

110-72-5
88.15
SCZVXVGZMZRGRU-UHFFFAOYSA-N
66071
CCNCCN
C4H12N2
MFCD00008166
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
N'-ethylethane-1,2-diamine

Specifications

110-72-5
100.0
128.0°C to 130.0°C
Authentic
C4H12N2
C2H5NHCH2CH2NH2
04, II, 690
n-ethylethylenediamine, 1,2-ethanediamine, n-ethyl, 2-aminoethyl ethyl amine, 2-ethylaminoethylamine, ethylenediamine, n-ethyl, ethylaminoethylamine, unii-941mvd708n, 1,2-ethanediamine, n1-ethyl, n-ethyl ethylenediamine
CCNCCN
88.15
88.15
N-Ethylethylenediamine
98.0
0.8300g/mL
10°C
98% min. (GC)
1.4380 to 1.4400
MFCD00008166
0.83
SCZVXVGZMZRGRU-UHFFFAOYSA-N
N'-ethylethane-1,2-diamine
66071
98+%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Highly flammable liquid and vapor.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 203-795-4

SDS
Documents

Documents

RUO – Research Use Only