missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcyclohexylamine 99.0+%, TCI America™

Catalog No. E050625ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E050625ML 25 mL
E0506500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E050625ML Supplier TCI America Supplier No. E050625ML

Chemical Identifiers

CAS 5459-93-8
Molecular Formula C8H17N
Molecular Weight (g/mol) 127.231
MDL Number MFCD00003834
InChI Key AGVKXDPPPSLISR-UHFFFAOYSA-N
Synonym n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e
PubChem CID 21609
IUPAC Name N-ethylcyclohexanamine
SMILES CCNC1CCCCC1

Specifications

Color Yellow
Boiling Point 164°C
Quantity 25 mL
UN Number 2734
Formula Weight 127.23
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material N-Ethylcyclohexylamine
RTECSNumber GX1225000
TSCA Yes