Learn More
N-Ethylcyclohexylamine, 97%
CAS: 5459-93-8 | C8H17N | 127.231 g/mol
Supplier: Thermo Scientific Chemicals L14221AE
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
| N-Ethylcyclohexylamine | |
| 0.85 | |
| 45°C (113°F) | |
| C8H17N | |
| MFCD00003834 | |
| UN2734 | |
| Air Sensitive | |
| AGVKXDPPPSLISR-UHFFFAOYSA-N | |
| N-ethylcyclohexanamine | |
| 21609 | |
| 97% |
| 5459-93-8 | |
| 163°C to 166°C | |
| Amine-like | |
| 1.452 | |
| 100 mL | |
| 2070280 | |
| n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e | |
| CCNC1CCCCC1 | |
| 127.231 | |
| 127.23 |
Chemical Identifiers
| 5459-93-8 | |
| 127.231 | |
| AGVKXDPPPSLISR-UHFFFAOYSA-N | |
| 21609 | |
| CCNC1CCCCC1 |
| C8H17N | |
| MFCD00003834 | |
| n-ethylcyclohexylamine, cyclohexanamine, n-ethyl, n-cyclohexylethylamine, vulkacit hx, accelerator hx, cyclohexylamine, n-ethyl, ethylamino cyclohexane, cyclohexyl ethyl amine, n-ethyl cyclohexyl amine, unii-yjk13p0h3e | |
| N-ethylcyclohexanamine |
Safety and Handling
GHS H Statement
H301-H311-H314-H318-H226-H332
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Flammable liquid and vapor.
Harmful if inhaled.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
H226-H302+H312+H332-H314-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, FLAMMABLE, N.O.S.
EINECSNumber : 226-733-8
RTECSNumber : GX1225000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only