missing translation for 'onlineSavingsMsg'
Learn More

N-Ethylcarbazole-3-carboxaldehyde 98.0+%, TCI America™

Catalog No. E03595G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E03595G 5 g
E035925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E03595G Supplier TCI America Supplier No. E03595G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 7570-45-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00004963
InChI Key QGJXVBICNCIWEL-UHFFFAOYSA-N
Synonym 9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
PubChem CID 82055
IUPAC Name 9-ethyl-9H-carbazole-3-carbaldehyde
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2

Specifications

Melting Point 90°C
Color Yellow
Boiling Point 255°C
Quantity 5 g
Formula Weight 223.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethylcarbazole-3-carboxaldehyde
EINECSNumber (5)-5798
TSCA No