missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethylacetamide 99.0+%, TCI America™
Supplier: TCI America E032425ML
Specifications
| N-Ethylacetamide | |
| -32°C | |
| 206°C | |
| MFCD00009029 | |
| acetamide, n-ethyl, acetamidoethane, acetoethylamide, ethylacetamide, n-aethylacetamid, n-acetylethylamine, n-aethylacetamid german, n-ethyl-acetamide, unii-m9y95ze6j9, 1-acetamidoethyl | |
| CCNC(C)=O | |
| 87.12 | |
| CHEBI:87370 | |
| ≥99.0% (GC) |
| 625-50-3 | |
| Yellow | |
| C4H9NO | |
| 25 mL | |
| PMDCZENCAXMSOU-UHFFFAOYSA-N | |
| N-ethylacetamide | |
| 12253 | |
| 87.12 | |
| Liquid |
Chemical Identifiers
| 625-50-3 | |
| 87.12 | |
| PMDCZENCAXMSOU-UHFFFAOYSA-N | |
| 12253 | |
| N-ethylacetamide |
| C4H9NO | |
| MFCD00009029 | |
| acetamide, n-ethyl, acetamidoethane, acetoethylamide, ethylacetamide, n-aethylacetamid, n-acetylethylamine, n-aethylacetamid german, n-ethyl-acetamide, unii-m9y95ze6j9, 1-acetamidoethyl | |
| CHEBI:87370 | |
| CCNC(C)=O |
Safety and Handling
EINECSNumber : (2)-3165
RTECSNumber : AB8500000
TSCA : Yes