missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-menthane-3-carboxamide 98.0+%, TCI America™

Catalog No. p-7129155
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E07965G 5 g
E079625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E07965G Supplier TCI America Supplier No. E07965G
Only null left

Chemical Identifiers

CAS 39711-79-0
Molecular Formula C13H25NO
Molecular Weight (g/mol) 211.35
MDL Number MFCD00130071
InChI Key VUNOFAIHSALQQH-UHFFFAOYNA-N
Synonym n-ethyl-p-menthane-3-carboxamide, ethyl menthane carboxamide, n-ethyl-2-isopropyl-5-methylcyclohexanecarboxamide, ws-3, n-ethyl-4-menthane-3-carboxamide, cyclohexanecarboxamide, n-ethyl-5-methyl-2-1-methylethyl, fema no. 3455, ws 3, n-ethyl-2-isopropyl-5-methylcyclohexane carboxamide
PubChem CID 62907
IUPAC Name N-ethyl-5-methyl-2-(propan-2-yl)cyclohexane-1-carboxamide
SMILES CCNC(=O)C1CC(C)CCC1C(C)C

Specifications

Melting Point 99°C
Color White
Quantity 5 g
Formula Weight 211.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethyl-p-menthane-3-carboxamide
EINECSNumber (3)-4186
TSCA Yes