missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Ethyl-p-menthane-3-carboxamide 98.0+%, TCI America™
Spécifications
Spécifications
| Melting Point | 99°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 211.35 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | N-Ethyl-p-menthane-3-carboxamide |
Sécurité et manipulation
| EINECSNumber | (3)-4186 |
| TSCA | Yes |