missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-3-carbazolecarboxaldehyde, 94%, Tech.

Catalog No. AC166030250
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC166030250 25 g Glass bottle
AC166031000 100 g Glass bottle
AC166035000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC166030250 Supplier Thermo Scientific Chemicals Supplier No. 166030250

CAS: 7570-45-8 | C15H13NO | 223.28 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 7570-45-8
Molecular Formula C15H13NO
Molecular Weight (g/mol) 223.28
MDL Number MFCD00004963
InChI Key QGJXVBICNCIWEL-UHFFFAOYSA-N
Synonym 9-ethyl-9h-carbazole-3-carbaldehyde, n-ethyl-3-carbazolecarboxaldehyde, n-ethylcarbazole-3-carboxaldehyde, 3-formyl-n-ethylcarbazole, n-ethylcarbazole-3-carbaldehyde, 9-ethyl-3-carbazolecarboxaldehyde, ccris 9052, n-ethyl carbazole-3-aldehyde, n-ethyl-carbazole-3-aldehyde, pubchem9556
PubChem CID 82055
IUPAC Name 9-ethylcarbazole-3-carbaldehyde
SMILES CCN1C2=C(C=CC=C2)C2=C1C=CC(C=O)=C2

Specifications

CAS Min % 92.0
CAS Max % 100.0
Melting Point 85°C to 91°C
Color Yellow to Ochre
Infrared Spectrum Authentic
Assay Percent Range 92% min. (HPLC)
Packaging Glass bottle
Quantity 25 g
Formula Weight 223.27
Percent Purity 94%
Grade Technical
Physical Form Crystalline Powder or Granules
Chemical Name or Material N-Ethyl-3-carbazolecarboxaldehyde
Show More Show Less
EINECSNumber 231-471-2

RUO – Research Use Only