missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-1-naphthylamine Hydrobromide 98.0+%, TCI America™

Catalog No. p-7128864
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E01495G 5 g
E014925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E01495G Supplier TCI America Supplier No. E01495G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 36966-04-8
Molecular Formula C12H14BrN
Molecular Weight (g/mol) 252.16
MDL Number MFCD00060154
InChI Key SBGSNWFFTCHTQG-UHFFFAOYSA-N
PubChem CID 44148659
IUPAC Name N-ethylnaphthalen-1-aminium bromide
SMILES [Br-].CC[NH2+]C1=C2C=CC=CC2=CC=C1

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 252.16
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethyl-1-naphthylamine Hydrobromide
EINECSNumber (4)-0349
TSCA No