missing translation for 'onlineSavingsMsg'
Learn More

N-Ethoxycarbonylphthalimide 98.0+%, TCI America™

Catalog No. p-7124559
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C068325G 25 g
C0683100G 100 g
C0683500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C068325G Supplier TCI America Supplier No. C068325G
Only null left

Chemical Identifiers

CAS 22509-74-6
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00005893
InChI Key VRHAQNTWKSVEEC-UHFFFAOYSA-N
Synonym n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
PubChem CID 31187
IUPAC Name ethyl 1,3-dioxoisoindole-2-carboxylate
SMILES CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 87°C
Color White
Quantity 25 g
Formula Weight 219.20
Percent Purity ≥98.0% (N)
Physical Form Crystalline Powder
Chemical Name or Material N-Ethoxycarbonylphthalimide [for Peptide Synthesis]
RTECSNumber NR3459500
TSCA Yes