missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)phthalimide, 97%

Catalog No. p-7022180
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1482214 25 g
AAA1482222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1482214 Supplier Thermo Scientific Chemicals Supplier No. A1482214

CAS: 22509-74-6 | C11H9NO4 | 219.196 g/mol

N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Ethoxycarbonyl)phthalimide is used to protect amine functional groups. Reagent for the conversion of amines and amino acids to their phthaloyl derivatives under mild conditions

Solubility
Insoluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 22509-74-6
Molecular Formula C11H9NO4
Molecular Weight (g/mol) 219.196
MDL Number MFCD00005893
InChI Key VRHAQNTWKSVEEC-UHFFFAOYSA-N
Synonym n-carbethoxyphthalimide, n-ethoxycarbonylphthalimide, n-carboethoxyphthalimide, phthalimide-n-carbethoxy, ethyl n-phthaloylcarbamate, n-ethoxycarbonyl phthalimide, 2h-isoindole-2-carboxylic acid, 1,3-dihydro-1,3-dioxo-, ethyl ester, ethyl 1,3-dioxoisoindoline-2-carboxylate, n-karbetoksi-ftalimid, 2-isoindolinecarboxylic acid, 1,3-dioxo-, ethyl ester
PubChem CID 31187
IUPAC Name ethyl 1,3-dioxoisoindole-2-carboxylate
SMILES CCOC(=O)N1C(=O)C2=CC=CC=C2C1=O

Specifications

Melting Point 78°C to 84°C
Quantity 25 g
Beilstein 196340
Solubility Information Insoluble in water.
Formula Weight 219.2
Percent Purity 97%
Chemical Name or Material N-(Ethoxycarbonyl)phthalimide
Hazard Category H302-H500
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 245-048-5
RTECSNumber NR3459500
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only