missing translation for 'onlineSavingsMsg'
Learn More

N-(Ethoxycarbonyl)nortropinone, 99%, Thermo Scientific™

Catalog No. p-7046296
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1384906 5 g
AAL1384914 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1384906 Supplier Thermo Scientific Chemicals Supplier No. L1384906
Only null left

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 32499-64-2
Molecular Formula C10H15NO3
Molecular Weight (g/mol) 197.234
MDL Number MFCD00078171
InChI Key ANEJUHJDPGTVIO-UHFFFAOYSA-N
Synonym ethyl 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylate, n-ethoxycarbonyl nortropinone, n-carbethoxy-4-nortropinone, n-carbethoxy-4-tropinone, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octan-3-one, 3-oxo-8-azabicyclo 3.2.1 octane-8-carboxylic acid ethyl ester, 8-azabicyclo 3.2.1 octane-8-carboxylic acid, 3-oxo-, ethyl ester, 8-ethoxycarbonyl-8-azabicyclo 3.2.1 octane-3-one, n-carbethoxynortropinone
PubChem CID 401882
IUPAC Name ethyl 3-oxo-8-azabicyclo[3.2.1]octane-8-carboxylate
SMILES CCOC(=O)N1C2CCC1CC(=O)C2

Specifications

Boiling Point 152°C to 153°C (11 mmHg)
Refractive Index 1.4895
Quantity 5 g
Beilstein 1530944
Formula Weight 197.23
Percent Purity 99%
Chemical Name or Material N-(Ethoxycarbonyl)nortropinone
TSCA No
Recommended Storage Ambient temperatures