missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%
CAS: 92122-45-7 | C26H32N2O6 | 468.55 g/mol
$113.86 - $363.66
Chemical Identifiers
| CAS | 92122-45-7 |
|---|---|
| Molecular Formula | C26H32N2O6 |
| Molecular Weight (g/mol) | 468.55 |
| MDL Number | MFCD00065660 |
| InChI Key | UMRUUWFGLGNQLI-ANBDAQEENA-N |
| Synonym | fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 |
| PubChem CID | 13585941 |
| IUPAC Name | (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
| SMILES | CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH2830103
|
Thermo Scientific Chemicals
H2830103 |
1 g |
Each for $113.86
|
|
|||||
|
AAH2830106
|
Thermo Scientific Chemicals
H2830106 |
5 g |
Each for $363.66
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 92122-45-7 | |
| 468.55 | |
| UMRUUWFGLGNQLI-ANBDAQEENA-N | |
| 13585941 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O |
| C26H32N2O6 | |
| MFCD00065660 | |
| fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid |
Specifications
| 135°C to 139°C | |
| C26H32N2O6 | |
| fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330 | |
| CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O | |
| 1 g | |
| 13585941 | |
| 468.54 | |
| N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine |
| 92122-45-7 | |
| MFCD00065660 | |
| UMRUUWFGLGNQLI-ANBDAQEENA-N | |
| (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid | |
| 468.55 | |
| +11° (c=1 in DMF) | |
| 98% |
Safety and Handling
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only