missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine, 98%

Catalog No. AAH2830103
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2830103 1 g
AAH2830106 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2830103 Supplier Thermo Scientific Chemicals Supplier No. H2830103
Only null left

CAS: 92122-45-7 | C26H32N2O6 | 468.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 92122-45-7
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.55
MDL Number MFCD00065660
InChI Key UMRUUWFGLGNQLI-ANBDAQEENA-N
Synonym fmoc-d-lys boc-oh, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-d-lysine, r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-yl methoxy carbonyl amino hexanoic acid, nepsilon-boc-nalpha-fmoc-d-lysine, r-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid, n epsilon-tert-butoxycarbonyl-n alpha-9-fluorenylmethoxycarbonyl-d-lysine, n-alpha-9-fluorenylmethyloxycarbonyl-n-epsilon-t-butyl-oxycarbonyl-d-lysine, fmocdlys boc oh, 2r-6-tert-butoxycarbonyl amino-2-9h-fluoren-9-ylmethoxy carbonyl amino hexanoic acid, ambotzfaa1330
PubChem CID 13585941
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCC[C@@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 135°C to 139°C
Quantity 1 g
Formula Weight 468.54
Optical Rotation +11° (c=1 in DMF)
Percent Purity 98%
Chemical Name or Material N(epsilon)-Boc-N(alpha)-Fmoc-D-lysine
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only