missing translation for 'onlineSavingsMsg'
Learn More

N(^e)-Benzyloxycarbonyl-L-lysine, 98%

Catalog No. AAA1602214
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1602214 25 g
AAA1602206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1602214 Supplier Thermo Scientific Chemicals Supplier No. A1602214

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 1155-64-2 | C14H20N2O4 | 280.32 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1155-64-2
Molecular Formula C14H20N2O4
Molecular Weight (g/mol) 280.32
MDL Number MFCD00002638
InChI Key CKGCFBNYQJDIGS-LBPRGKRZSA-N
Synonym h-lys z-oh, n6-cbz-l-lysine, n-epsilon-carbobenzyloxy-l-lysine, n epsilon-benzyloxycarbonyl-l-lysine, n6-benzyloxycarbonyl-l-lysine, nepsilon-carbobenzoxy-l-lysine, nepsilon-carbobenzyloxy-l-lysine, l-lys cbz-oh, 2s-2-amino-6-benzyloxy carbonyl amino hexanoic acid, 6-n-cbz-l-lysine
PubChem CID 1715626
IUPAC Name (2S)-2-amino-6-(phenylmethoxycarbonylamino)hexanoic acid
SMILES N[C@@H](CCCCNC(=O)OCC1=CC=CC=C1)C(O)=O

Specifications

Melting Point 259°C (decomposition)
Beilstein 2222482
Quantity 25 g
Formula Weight 280.32
Percent Purity 98%
Chemical Name or Material N(epsilon)-Benzyloxycarbonyl-L-lysine
EINECSNumber 214-585-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only