missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyltriethoxysilane, 95%

Catalog No. AAL1729314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1729314 25 g
AAL1729322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1729314 Supplier Thermo Scientific Chemicals Supplier No. L1729314
Only null left

CAS: 18536-91-9 | C18H40O3Si | 332.6 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 18536-91-9
Molecular Formula C18H40O3Si
Molecular Weight (g/mol) 332.6
MDL Number MFCD00048437
InChI Key YGUFXEJWPRRAEK-UHFFFAOYSA-N
Synonym silane, dodecyltriethoxy, n-dodecyltriethoxysilane, lauryl triethoxy silane, dodecyl triethoxy silane, acmc-209emw, silane,dodecyltriethoxy, dodecyltriethoxysilane, technical
PubChem CID 87696
IUPAC Name dodecyl(triethoxy)silane
SMILES CCCCCCCCCCCC[Si](OCC)(OCC)OCC

Specifications

Boiling Point 152°C to 153°C (3mmHg)
Quantity 25 g
Linear Formula CH3(CH2)11Si(OCH2CH3)3
Beilstein 2414223
Flash Point >110°C (230°F)
Formula Weight 332.6
Percent Purity 95%
Refractive Index 1.433
Sensitivity Moisture sensitive
Density 0.89
Chemical Name or Material n-Dodecyltriethoxysilane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 242-409-9
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only