missing translation for 'onlineSavingsMsg'
Learn More

N-Dodecylmethylamine, 98%

Catalog No. AAL0702506
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0702506 5 g
AAL0702503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0702506 Supplier Thermo Scientific Chemicals Supplier No. L0702506

CAS: 7311-30-0 | C13H29N | 199.382 g/mol

N-Dodecylmethylamine is used as intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Dodecylmethylamine is used as intermediates.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.

Chemical Identifiers

CAS 7311-30-0
Molecular Formula C13H29N
Molecular Weight (g/mol) 199.382
MDL Number MFCD00026574
InChI Key OMEMQVZNTDHENJ-UHFFFAOYSA-N
Synonym n-methyldodecylamine, dodecyl methyl amine, dodecylmethylamine, 1-dodecanamine, n-methyl, n-dodecylmethylamine, dodecyl-methyl-amine, unii-88nn43i94s, methyldodecylamine, n-methyldodecanamine, n-methyl-n-dodecylamine
PubChem CID 81746
IUPAC Name N-methyldodecan-1-amine
SMILES CCCCCCCCCCCCNC

Specifications

Density 0.795
Boiling Point 90°C to 92°C (0.9 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4395
Quantity 5 g
UN Number UN2735
Beilstein 1739959
Solubility Information Not miscible in water.
Formula Weight 199.38
Percent Purity 98%
Chemical Name or Material N-Dodecylmethylamine
Show More Show Less
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber 230-771-0
TSCA No

RUO – Research Use Only