Learn More
n-Dodecylbenzene, 97%
Description
n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, nitric acid.
Specifications
Specifications
| Melting Point | -7°C |
| Density | 0.856 |
| Boiling Point | 331°C |
| Flash Point | 124°C (255°F) |
| Refractive Index | 1.482 |
| Quantity | 5 g |
| Beilstein | 1909107 |
| Solubility Information | Insoluble in water. |
| Formula Weight | 246.44 |
| Percent Purity | 97% |
| Show More |
Safety and Handling
| EINECSNumber | 204-591-8 |
| RTECSNumber | CZ9540000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only