missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecylbenzene, 97%

Catalog No. AAB2136206
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2136206 5 g
AAB2136214 25 g
AAB2136222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2136206 Supplier Thermo Scientific Chemicals Supplier No. B2136206

CAS: 123-01-3 | C18H30 | 246.438 g/mol

n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Dodecylbenzene, is a precursor to sodium dodecylbenzenesulfonate, a surfactant that is a key ingredient of household laundry detergents, such as Tide. It is a widely used cleaner, dyeing assistant in textile industrial, degrease agent in electroplate, leather, de- inking agent in papering, rubber and plastic, break emulsion agent in oil industry, artesian well foaming agent in oil well.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents, nitric acid.

Chemical Identifiers

CAS 123-01-3
Molecular Formula C18H30
Molecular Weight (g/mol) 246.438
MDL Number MFCD00008974
InChI Key KWKXNDCHNDYVRT-UHFFFAOYSA-N
Synonym 1-phenyldodecane, benzene, dodecyl, n-dodecylbenzene, laurylbenzene, phenyldodecane, marlican, dodecane, 1-phenyl, detergent alkylate, alkylate p 1, phenyldodecan
PubChem CID 31237
IUPAC Name dodecylbenzene
SMILES CCCCCCCCCCCCC1=CC=CC=C1

Specifications

Melting Point -7°C
Density 0.856
Boiling Point 331°C
Flash Point 124°C (255°F)
Refractive Index 1.482
Quantity 5 g
Beilstein 1909107
Solubility Information Insoluble in water.
Formula Weight 246.44
Percent Purity 97%
Chemical Name or Material n-Dodecylbenzene
Show More Show Less
EINECSNumber 204-591-8
RTECSNumber CZ9540000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only