missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate
Description
For protein solubilization
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | White |
| Flash Point | 110°C (230°F) |
| Quantity | 5 g |
| Beilstein | 4145308 |
| Sensitivity | Hygroscopic |
| Solubility Information | Soluble at 1M in water. |
| Formula Weight | 335.55 |
| Physical Form | Powder |
| Chemical Name or Material | N-Dodecyl-N,N-dimethyl-3-ammonio-1-propanesulfonate |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H302-H312-H332-H315-H319-H335 Harmful if swallowed. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 239-002-3 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only