missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Dodecyl-beta-D-maltoside, 99%, high purity
CAS: 69227-93-6 | C24H46O11 | 510.6 g/mol
Supplier: Thermo Scientific Chemicals 329370050
| Quantity | 5 g |
|---|---|
| Packaging | Glass bottle |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 69227-93-6 | |
| 510.6 | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| 114880 | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol |
| C24H46O11 | |
| MFCD00043012 | |
| n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
| CHEBI:43769 | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O |
Specifications
| n-Dodecyl-β-D-maltoside | |
| 98.5 | |
| 217.0°C | |
| Authentic | |
| Glass bottle | |
| 5 g | |
| 17, V,8, 12 | |
| + 47.50 | |
| Solubility in water: soluble (100g/L). Other solubilities: soluble in methanol, ethanol | |
| CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O | |
| 510.6 | |
| CHEBI:43769 | |
| 99% | |
| Powder |
| 69227-93-6 | |
| 100.0 | |
| White | |
| 98.5% min. (ELSD) (HPLC) | |
| C24H46O11 | |
| MFCD00043012 | |
| +47.50 (20.00°C c=1,water) | |
| n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside | |
| NLEBIOOXCVAHBD-QKMCSOCLSA-N | |
| (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol | |
| 114880 | |
| 510.6 | |
| High Purity |