missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl-beta-D-maltopyranoside

Catalog No. AAJ6686906
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ6686906 5 g
AAJ66869ME 500 mg
AAJ6686903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAJ6686906 Supplier Thermo Scientific Chemicals Supplier No. J6686906
Only null left

A non-ionic detergent useful in membrane solubilization studies | CAS: 69227-93-6 | C24H46O11 | 510.621 g/mol

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

Solubility
Soluble in water at 10% w/v

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.

Chemical Identifiers

CAS 69227-93-6
Molecular Formula C24H46O11
Molecular Weight (g/mol) 510.621
MDL Number MFCD00043012
InChI Key NLEBIOOXCVAHBD-QKMCSOCLSA-N
Synonym n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
PubChem CID 114880
ChEBI CHEBI:43769
IUPAC Name (2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

Melting Point 224°C to 226°C
Color White
Quantity 5 g
Beilstein 55318
Solubility Information Soluble in water at 10% w/v
Formula Weight 510.62
Physical Form Powder
Chemical Name or Material n-Dodecyl-beta-D-maltopyranoside
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only