missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecyl-beta-D-maltopyranoside

A non-ionic detergent useful in membrane solubilization studies | CAS: 69227-93-6 | C24H46O11 | 510.621 g/mol

Supplier:  Thermo Scientific Chemicals J6686903

Quantity 1 g

 View more versions of this product

Catalog No. AAJ6686903


Only null left

Description

Description

N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-dodecyl-β-D-maltoside is used as a non-ionic detergent for stabilization and activation of enzymes and for membrane research. It has also been used for the isolation of the mitochondrial cytochrome c oxidize.

Solubility
Soluble in water at 10% w/v

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications

Chemical Identifiers

69227-93-6
510.621
NLEBIOOXCVAHBD-QKMCSOCLSA-N
114880
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
C24H46O11
MFCD00043012
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
CHEBI:43769
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O

Specifications

n-Dodecyl-beta-D-maltopyranoside
224°C to 226°C
C24H46O11
MFCD00043012
n-dodecyl-beta-d-maltoside, lauryl maltoside, dodecyl maltoside, dodecyl-beta-d-maltoside, n-dodecyl b-d-maltoside, unii-di107e57b4, lauryl-beta-d-maltoside, dodecyl beta-d-maltoside, dodecyl b-d-maltopyranoside, dodecyl beta-d-maltopyranoside
NLEBIOOXCVAHBD-QKMCSOCLSA-N
(2R,3R,4S,5S,6R)-2-[(2R,3S,4R,5R,6R)-6-dodecoxy-4,5-dihydroxy-2-(hydroxymethyl)oxan-3-yl]oxy-6-(hydroxymethyl)oxane-3,4,5-triol
114880
510.62
69227-93-6
White
1 g
55318
Soluble in water at 10% w/v
CCCCCCCCCCCCOC1C(C(C(C(O1)CO)OC2C(C(C(C(O2)CO)O)O)O)O)O
510.621
CHEBI:43769
Powder
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Store at -20°C

SDS
Documents

Documents

Promotions

Promotions