missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ n-Dodecanoylsucrose, Calbiochem™,

Catalog No. 3243745GM
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
3243745GM 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 3243745GM Supplier MilliporeSigma™ Supplier No. 3243745GM

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 25339-99-5
Molecular Formula C24H44O12
Synonym n-Monododecanoate-α-D-glucopyranoside, β-D-Fructofuranosylsucrose Monolaurate

Specifications

Color White
Quantity 5 g
Packaging Glass Bottle