missing translation for 'onlineSavingsMsg'
Learn More

n-Dodecane, 99%, anhydrous, AcroSeal™

Catalog No. p-4258987
Change view
Click to view available options
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC434591000 100 mL AcroSeal™ Glass Bottle
AC434590010 1 L AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC434591000 Supplier Thermo Scientific Chemicals Supplier No. 434591000
Only null left

CAS: 112-40-3 | C12H26 | 170.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 112-40-3
Molecular Formula C12H26
Molecular Weight (g/mol) 170.34
InChI Key SNRUBQQJIBEYMU-UHFFFAOYSA-N
Synonym n-dodecane, dihexyl, bihexyl, adakane 12, n-dodecan, n-dodecan german, duodecane, alkanes, c10-14, undecane, methyl, ccris 661
PubChem CID 8182
ChEBI CHEBI:28817
IUPAC Name dodecane
SMILES CCCCCCCCCCCC

Specifications

Melting Point -12.0°C
Color Colorless
Density 0.7530g/mL
Boiling Point 215.0°C to 217.0°C
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging AcroSeal™ Glass Bottle
Refractive Index 1.4210 to 1.4230
Linear Formula CH3(CH2)10CH3
Quantity 100 mL
Specific Gravity 0.753
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, acetone and ether
Viscosity <7 mm2/s (40°C)
Formula Weight 170.34
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material n-Dodecane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
May be fatal if swallowed and enters airways.
Repeated exposure may cause skin dryness or cracking.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTRE or doctor/physician.
EINECSNumber 203-967-9

RUO – Research Use Only