missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine ethyl ester, 98%

CAS: 69555-14-2 | C17H17NO2 | 267.328 g/mol

$143.70 - $490.75

Chemical Identifiers

CAS 69555-14-2
Molecular Formula C17H17NO2
Molecular Weight (g/mol) 267.328
MDL Number MFCD00010590
InChI Key QUGJYNGNUBHTNS-UHFFFAOYSA-N
Synonym ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
PubChem CID 319508
IUPAC Name ethyl 2-(benzhydrylideneamino)acetate
SMILES CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2558006
View Documents
Thermo Scientific Chemicals
B2558006
5 g N/A
Only null left
 
AAB2558014
View Documents
Thermo Scientific Chemicals
B2558014
25 g N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

69555-14-2
267.328
QUGJYNGNUBHTNS-UHFFFAOYSA-N
319508
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
C17H17NO2
MFCD00010590
ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
ethyl 2-(benzhydrylideneamino)acetate

Specifications

51°C to 55°C
C17H17NO2
ethyl n-diphenylmethylene glycinate, n-diphenylmethylene glycine ethyl ester, ethyl 2-diphenylmethylene amino acetate, ethyl 2-diphenylmethyleneamino acetate, diphenylmethylene-glycine ethyl ester, ethyl 2-benzhydrylideneamino acetate, ethyl 2-diphenylmethylidene amino acetate, diphenylmethylene glycine ethylester, ethyl diphenylmethylenamino acetate, diphenylmethylene glycine ethyl ester
CCOC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2
5 g
>110°C (230°F)
267.33
98%
69555-14-2
MFCD00010590
QUGJYNGNUBHTNS-UHFFFAOYSA-N
ethyl 2-(benzhydrylideneamino)acetate
267.328
319508
Moisture sensitive
N-(Diphenylmethylene)glycine ethyl ester
Videos
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only