missing translation for 'onlineSavingsMsg'
Learn More

N-(Diethylcarbamoyl)-N-methoxyformamide 98.0+%, TCI America™

Catalog No. D23441G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23441G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D23441G Supplier TCI America Supplier No. D23441G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 146039-03-4
Molecular Formula C7H14N2O3
Molecular Weight (g/mol) 174.2
MDL Number MFCD00191345
InChI Key ABUWPGZRKFDPIX-UHFFFAOYSA-N
Synonym N,N-Diethyl-N′C-formyl-N′C-methoxyurea
PubChem CID 567783
IUPAC Name N-(diethylcarbamoyl)-N-methoxyformamide
SMILES CCN(CC)C(=O)N(C=O)OC

Specifications

Color Yellow
Boiling Point 96°C
Quantity 1 g
Formula Weight 174.20
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material N-(Diethylcarbamoyl)-N-methoxyformamide [Selective Formylating Reagent]