missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Decyltrimethoxysilane, 97%
Description
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.
Solubility
Soluble in organic solvents. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.
Specifications
Specifications
| Boiling Point | 102°C to 106°C (3.5mmHg) |
| Melting Point | -37°C |
| Quantity | 100 g |
| Flash Point | 117°C (242°F) |
| Refractive Index | 1.422 |
| Sensitivity | Moisture sensitive |
| Density | 0.9 |
| Beilstein | 8625082 |
| Solubility Information | Soluble in organic solvents. Insoluble in water. |
| Formula Weight | 262.47 |
| Show More |
Safety and Handling
| Hazard Category | H315-H319 |
| Hazard Statement | GHS H Statement H315-H319 Causes skin irritation. Causes serious eye irritation. |
| Precautionary Statement | P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362 |
| EINECSNumber | 000-000-0 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only