missing translation for 'onlineSavingsMsg'
Learn More

n-Decyltrimethoxysilane, 97%

Catalog No. AAL1696614
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1696614 25 g
AAL1696622 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1696614 Supplier Thermo Scientific Chemicals Supplier No. L1696614
Only null left

CAS: 5575-48-4 | C13H30O3Si | 262.47 g/mol

n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Decyltrimethoxysilane is used as a coupling agent and as a pharmaceutical intermediate.

Solubility
Soluble in organic solvents. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store under dry inert gas. It is sensitive to moisture. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5575-48-4
Molecular Formula C13H30O3Si
Molecular Weight (g/mol) 262.47
MDL Number MFCD00069135
InChI Key KQAHMVLQCSALSX-UHFFFAOYSA-N
Synonym n-decyltrimethoxysilane, decyltrimethoxysilan, trimethoxydecylsilane, decyl trimethoxy silane, silane,decyltrimethoxy, decyl-trimethoxysilane
PubChem CID 2758023
IUPAC Name decyl(trimethoxy)silane
SMILES CCCCCCCCCC[Si](OC)(OC)OC

Specifications

Boiling Point 102°C to 106°C (3.5mmHg)
Melting Point -37°C
Quantity 25 g
Beilstein 8625082
Flash Point 117°C (242°F)
Solubility Information Soluble in organic solvents. Insoluble in water.
Formula Weight 262.47
Percent Purity 98%
Refractive Index 1.422
Sensitivity Moisture sensitive
Density 0.9
Chemical Name or Material n-Decyltrimethoxysilane
Show More Show Less
Hazard Category H315-H319
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only